Anti-OSMR Mouse Monoclonal Antibody [clone: 469221]

Supplier: R & D Systems
MAB4389
220020-874EA 633.42 USD
220020-874
Anti-OSMR Mouse Monoclonal Antibody [clone: 469221]
Antibodies
Type: Primary
Antigen: OSMR
Clonality: Monoclonal
Clone: 469221
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR