Anti-CD109 Mouse Monoclonal Antibody [clone: 496920]

Supplier: R & D Systems
MAB4385
102885-726EA 600.15 USD
102885-726
Anti-CD109 Mouse Monoclonal Antibody [clone: 496920]
Antibodies
Type: Primary
Antigen: CD109
Clonality: Monoclonal
Clone: 496920
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR