Anti-CTF1 Rat Monoclonal Antibody [clone: 91907]

Supplier: R & D Systems
MAB438
220020-873EA 763.57 USD
220020-873
Anti-CTF1 Rat Monoclonal Antibody [clone: 91907]
Antibodies
Type: Primary
Antigen: CTF1
Clonality: Monoclonal
Clone: 91907
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR