Anti-TSC1 Mouse Monoclonal Antibody [clone: 488915]

Supplier: R & D Systems
MAB4379
102885-724EA 594.37 USD
102885-724
Anti-TSC1 Mouse Monoclonal Antibody [clone: 488915]
Antibodies
Type: Primary
Antigen: TSC1
Clonality: Monoclonal
Clone: 488915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR