Anti-PRKCB Mouse Monoclonal Antibody [clone: 469832]

Supplier: R & D Systems
MAB4378
220020-872EA 577.02 USD
220020-872
Anti-PRKCB Mouse Monoclonal Antibody [clone: 469832]
Antibodies
Type: Primary
Antigen: PRKCB
Clonality: Monoclonal
Clone: 469832
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR