Anti-SP Mouse Monoclonal Antibody [clone: 266815]

Supplier: R & D Systems
MAB4375
220020-871EA 647.88 USD
220020-871
Anti-SP Mouse Monoclonal Antibody [clone: 266815]
Antibodies
Type: Primary
Antigen: SP
Clonality: Monoclonal
Clone: 266815
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR