Anti-C1QL2 Rat Monoclonal Antibody [clone: 384921]

Supplier: R & D Systems
MAB4374
220020-870EA 594.37 USD
220020-870
Anti-C1QL2 Rat Monoclonal Antibody [clone: 384921]
Antibodies
Type: Primary
Antigen: C1QL2
Clonality: Monoclonal
Clone: 384921
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR