Anti-NR3C2 Mouse Monoclonal Antibody [clone: 385707]

Supplier: R & D Systems
MAB4369
220020-869EA 577.02 USD
220020-869
Anti-NR3C2 Mouse Monoclonal Antibody [clone: 385707]
Antibodies
Type: Primary
Antigen: NR3C2
Clonality: Monoclonal
Clone: 385707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR