Anti-PRKCQ Mouse Monoclonal Antibody [clone: 453416]

Supplier: R & D Systems
MAB4368
220020-868EA 594.37 USD
220020-868
Anti-PRKCQ Mouse Monoclonal Antibody [clone: 453416]
Antibodies
Simple Western

Type: Primary
Antigen: PRKCQ
Clonality: Monoclonal
Clone: 453416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR