Anti-LAMTOR3 Mouse Monoclonal Antibody [clone: 448406]

Supplier: R & D Systems
MAB4367
220020-867EA 561.11 USD
220020-867
Anti-LAMTOR3 Mouse Monoclonal Antibody [clone: 448406]
Antibodies
Type: Primary
Antigen: LAMTOR3
Clonality: Monoclonal
Clone: 448406
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR