Anti-SYT1 Mouse Monoclonal Antibody [clone: ASV30]

Supplier: R & D Systems
MAB43641
220020-866EA 577.02 USD
220020-866
Anti-SYT1 Mouse Monoclonal Antibody [clone: ASV30]
Antibodies
Type: Primary
Antigen: SYT1
Clonality: Monoclonal
Clone: ASV30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR