Anti-SYT1 Mouse Monoclonal Antibody [clone: ASV48]

Supplier: R & D Systems
MAB4364
220020-865EA 577.02 USD
220020-865
Anti-SYT1 Mouse Monoclonal Antibody [clone: ASV48]
Antibodies
Type: Primary
Antigen: SYT1
Clonality: Monoclonal
Clone: ASV48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR