Anti-ERBB2 Mouse Monoclonal Antibody [clone: 416727]

Supplier: R & D Systems
MAB4360
220020-864EA 594.37 USD
220020-864
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 416727]
Antibodies
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 416727
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR