Anti-PINK1 Rat Monoclonal Antibody [clone: 420920]

Supplier: R & D Systems
MAB4357
220020-862EA 594.37 USD
220020-862
Anti-PINK1 Rat Monoclonal Antibody [clone: 420920]
Antibodies
Type: Primary
Antigen: PINK1
Clonality: Monoclonal
Clone: 420920
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR