Anti-FKBP1A Rat Monoclonal Antibody [clone: 422617]

Supplier: R & D Systems
MAB4356
220020-861EA 577.02 USD
220020-861
Anti-FKBP1A Rat Monoclonal Antibody [clone: 422617]
Antibodies
Type: Primary
Antigen Symbol: FKBP1A
Antigen Name: FK506 binding protein 1a
Antigen Synonyms: FKBP12, Fkbp2
Clonality: Monoclonal
Clone: 422617
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR