Anti-PIM2 Mouse Monoclonal Antibody [clone: 447023]

Supplier: R & D Systems
MAB4355
220020-860EA 561.11 USD
220020-860
Anti-PIM2 Mouse Monoclonal Antibody [clone: 447023]
Antibodies
Type: Primary
Antigen: PIM2
Clonality: Monoclonal
Clone: 447023
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR