Anti-CEACAM4 Mouse Monoclonal Antibody [clone: 568410]

Supplier: R & D Systems
MAB4354
102885-720EA 594.37 USD
102885-720
Anti-CEACAM4 Mouse Monoclonal Antibody [clone: 568410]
Antibodies
Type: Primary
Antigen: CEACAM4
Clonality: Monoclonal
Clone: 568410
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR