Anti-TICAM2 Rat Monoclonal Antibody [clone: 757712]

Supplier: R & D Systems
MAB43481
102885-718EA 577.02 USD
102885-718
Anti-TICAM2 Rat Monoclonal Antibody [clone: 757712]
Antibodies
Type: Primary
Antigen: TICAM2
Clonality: Monoclonal
Clone: 757712
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR