Anti-PON3 Rat Monoclonal Antibody [clone: 454929]

Supplier: R & D Systems
MAB4345
220020-858EA 561.11 USD
220020-858
Anti-PON3 Rat Monoclonal Antibody [clone: 454929]
Antibodies
Type: Primary
Antigen: PON3
Clonality: Monoclonal
Clone: 454929
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR