Anti-PON2 Rat Monoclonal Antibody [clone: 453709]

Supplier: R & D Systems
MAB4344
220020-857EA 561.11 USD
220020-857
Anti-PON2 Rat Monoclonal Antibody [clone: 453709]
Antibodies
Type: Primary
Antigen: PON2
Clonality: Monoclonal
Clone: 453709
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR