Anti-NUMB Mouse Monoclonal Antibody [clone: 447716]

Supplier: R & D Systems
MAB4338
220020-854EA 577.02 USD
220020-854
Anti-NUMB Mouse Monoclonal Antibody [clone: 447716]
Antibodies
Type: Primary
Antigen: NUMB
Clonality: Monoclonal
Clone: 447716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR