Anti-NETO1 Mouse Monoclonal Antibody [clone: 532509]

Supplier: R & D Systems
MAB4337
220020-853EA 561.11 USD
220020-853
Anti-NETO1 Mouse Monoclonal Antibody [clone: 532509]
Antibodies
Type: Primary
Antigen Symbol: NETO1
Antigen Name: neuropilin (NRP) and tolloid (TLL)-like 1
Antigen Synonyms: BCTL1, BTCL1
Clonality: Monoclonal
Clone: 532509
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR