Anti-S100 A13 Mouse Monoclonal Antibody [clone: 854114]

Supplier: R & D Systems
MAB4327
102885-714EA 561.11 USD
102885-714
Anti-S100 A13 Mouse Monoclonal Antibody [clone: 854114]
Antibodies
Type: Primary
Antigen: S100 A13
Clonality: Monoclonal
Clone: 854114
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR