Anti-ATG4A Mouse Monoclonal Antibody [clone: 514914]

Supplier: R & D Systems
MAB4324
220020-850EA 577.02 USD
220020-850
Anti-ATG4A Mouse Monoclonal Antibody [clone: 514914]
Antibodies
Type: Primary
Antigen: ATG4A
Clonality: Monoclonal
Clone: 514914
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR