Anti-LAMP1 Rat Monoclonal Antibody [clone: 747203]

Supplier: R & D Systems
MAB4320
102885-712EA 594.37 USD
102885-712
Anti-LAMP1 Rat Monoclonal Antibody [clone: 747203]
Antibodies
Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: 747203
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR