Anti-EPHB3 Rat Monoclonal Antibody [clone: 521002]

Supplier: R & D Systems
MAB432
220020-849EA 561.11 USD
220020-849
Anti-EPHB3 Rat Monoclonal Antibody [clone: 521002]
Antibodies
Type: Primary
Antigen: EPHB3
Clonality: Monoclonal
Clone: 521002
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR