Anti-COL25A1 Mouse Monoclonal Antibody [clone: 540802]

Supplier: R & D Systems
MAB4317
102885-710EA 577.02 USD
102885-710
Anti-COL25A1 Mouse Monoclonal Antibody [clone: 540802]
Antibodies
Type: Primary
Antigen: COL25A1
Clonality: Monoclonal
Clone: 540802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR