Anti-CRK Mouse Monoclonal Antibody [clone: 463414]

Supplier: R & D Systems
MAB4316
102885-708EA 577.02 USD
102885-708
Anti-CRK Mouse Monoclonal Antibody [clone: 463414]
Antibodies
Type: Primary
Antigen: CRK
Clonality: Monoclonal
Clone: 463414
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR