Anti-DLL3 Mouse Monoclonal Antibody [clone: 378703]

Supplier: R & D Systems
MAB4315
220020-848EA 655.11 USD
220020-848
Anti-DLL3 Mouse Monoclonal Antibody [clone: 378703]
Antibodies
Type: Primary
Antigen: DLL3
Clonality: Monoclonal
Clone: 378703
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR