Anti-CCND1 Mouse Monoclonal Antibody [clone: 328303]

Supplier: R & D Systems
MAB4314
220020-847EA 577.02 USD
220020-847
Anti-CCND1 Mouse Monoclonal Antibody [clone: 328303]
Antibodies
Type: Primary
Antigen: CCND1
Clonality: Monoclonal
Clone: 328303
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR