Anti-FSHB Mouse Monoclonal Antibody [clone: 405326]

Supplier: R & D Systems
MAB4310
220020-846EA 577.02 USD
220020-846
Anti-FSHB Mouse Monoclonal Antibody [clone: 405326]
Antibodies
Type: Primary
Antigen Symbol: FSHB
Antigen Name: follicle stimulating hormone, beta polypeptide
Antigen Synonyms: HH24
Clonality: Monoclonal
Clone: 405326
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR