Anti-NFKBIE Mouse Monoclonal Antibody [clone: 418515]

Supplier: R & D Systems
MAB4300
220020-843EA 577.02 USD
220020-843
Anti-NFKBIE Mouse Monoclonal Antibody [clone: 418515]
Antibodies
Type: Primary
Antigen Symbol: NFKBIE
Antigen Name: nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, epsilon
Antigen Synonyms: IKBE
Clonality: Monoclonal
Clone: 418515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR