Anti-IL22RA1 Rat Monoclonal Antibody [clone: 496504]

Supplier: R & D Systems
MAB4294
220020-839EA 594.37 USD
220020-839
Anti-IL22RA1 Rat Monoclonal Antibody [clone: 496504]
Antibodies
Type: Primary
Antigen: IL22RA1
Clonality: Monoclonal
Clone: 496504
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR