Anti-TPSB2 Rat Monoclonal Antibody [clone: 286828]

Supplier: R & D Systems
MAB4288
220020-838EA 779.48 USD
220020-838
Anti-TPSB2 Rat Monoclonal Antibody [clone: 286828]
Antibodies
Type: Primary
Antigen: TPSB2
Clonality: Monoclonal
Clone: 286828
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR