Anti-CXCR4 Mouse Monoclonal Antibody [clone: 374606]

Supplier: R & D Systems
MAB4287
220020-837EA 561.11 USD
220020-837
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 374606]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 374606
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR