Anti-GPBAR1 Mouse Monoclonal Antibody [clone: 409522]

Supplier: R & D Systems
MAB4286
220020-836EA 594.37 USD
220020-836
Anti-GPBAR1 Mouse Monoclonal Antibody [clone: 409522]
Antibodies
Type: Primary
Antigen Symbol: GPBAR1
Antigen Name: G protein-coupled bile acid receptor 1
Antigen Synonyms: BG37, GPCR19, GPR131, M-BAR, TGR5
Clonality: Monoclonal
Clone: 409522
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR