Anti-GLP2R Mouse Monoclonal Antibody [clone: 413801]

Supplier: R & D Systems
MAB4285
220020-835EA 594.37 USD
220020-835
Anti-GLP2R Mouse Monoclonal Antibody [clone: 413801]
Antibodies
Type: Primary
Antigen Symbol: GLP2R
Antigen Name: glucagon-like peptide 2 receptor
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 413801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR