Anti-CLDN12 Mouse Monoclonal Antibody [clone: 393314]

Supplier: R & D Systems
MAB4284
220020-834EA 594.37 USD
220020-834
Anti-CLDN12 Mouse Monoclonal Antibody [clone: 393314]
Antibodies
Type: Primary
Antigen: CLDN12
Clonality: Monoclonal
Clone: 393314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR