Anti-CCL12 Rat Monoclonal Antibody [clone: 147801]

Supplier: R & D Systems
MAB4281
102885-706EA 608.83 USD
102885-706
Anti-CCL12 Rat Monoclonal Antibody [clone: 147801]
Antibodies
Type: Primary
Antigen: CCL12
Clonality: Monoclonal
Clone: 147801
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR