Anti-CLDN10 Mouse Monoclonal Antibody [clone: 394513]

Supplier: R & D Systems
MAB4279
220020-831EA 594.37 USD
220020-831
Anti-CLDN10 Mouse Monoclonal Antibody [clone: 394513]
Antibodies
Type: Primary
Antigen: CLDN10
Clonality: Monoclonal
Clone: 394513
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR