Anti-REN1 Rat Monoclonal Antibody [clone: 434703]

Supplier: R & D Systems
MAB42771
220020-830EA 577.02 USD
220020-830
Anti-REN1 Rat Monoclonal Antibody [clone: 434703]
Antibodies
Type: Primary
Antigen: REN1
Clonality: Monoclonal
Clone: 434703
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR