Anti-SAA4 Rat Monoclonal Antibody [clone: 398104]

Supplier: R & D Systems
MAB4276
102885-704EA 561.11 USD
102885-704
Anti-SAA4 Rat Monoclonal Antibody [clone: 398104]
Antibodies
Type: Primary
Antigen: SAA4
Clonality: Monoclonal
Clone: 398104
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR