Anti-CXCL17 Rat Monoclonal Antibody [clone: 510614]

Supplier: R & D Systems
MAB4270
220020-828EA 594.37 USD
220020-828
Anti-CXCL17 Rat Monoclonal Antibody [clone: 510614]
Antibodies
Type: Primary
Antigen: CXCL17
Clonality: Monoclonal
Clone: 510614
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR