Anti-ILK Mouse Monoclonal Antibody [clone: 443208]

Supplier: R & D Systems
MAB4266
220020-827EA 594.37 USD
220020-827
Anti-ILK Mouse Monoclonal Antibody [clone: 443208]
Antibodies
Type: Primary
Antigen: ILK
Clonality: Monoclonal
Clone: 443208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR