Anti-TNFRSF1B Hamster Monoclonal Antibody [clone: TR7532]

Supplier: R & D Systems
MAB4261
220020-825EA 763.57 USD
220020-825
Anti-TNFRSF1B Hamster Monoclonal Antibody [clone: TR7532]
Antibodies
Type: Primary
Antigen: TNFRSF1B
Clonality: Monoclonal
Clone: TR7532
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR