Anti-JAK1 Mouse Monoclonal Antibody [clone: 452804]

Supplier: R & D Systems
MAB42601
220020-824EA 594.37 USD
220020-824
Anti-JAK1 Mouse Monoclonal Antibody [clone: 452804]
Antibodies
Type: Primary
Antigen: JAK1
Clonality: Monoclonal
Clone: 452804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR