Anti-PXN Mouse Monoclonal Antibody [clone: 492918]

Supplier: R & D Systems
MAB4259
220020-821EA 577.02 USD
220020-821
Anti-PXN Mouse Monoclonal Antibody [clone: 492918]
Antibodies
Type: Primary
Antigen: PXN
Clonality: Monoclonal
Clone: 492918
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR