Anti-TNFRSF1A Rat Monoclonal Antibody [clone: 47803]

Supplier: R & D Systems
MAB425
220020-819EA 779.48 USD
220020-819
Anti-TNFRSF1A Rat Monoclonal Antibody [clone: 47803]
Antibodies
Type: Primary
Antigen: TNFRSF1A
Clonality: Monoclonal
Clone: 47803
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR