Anti-GP5 Mouse Monoclonal Antibody [clone: 508209]

Supplier: R & D Systems
MAB4249
220020-817EA 594.37 USD
220020-817
Anti-GP5 Mouse Monoclonal Antibody [clone: 508209]
Antibodies
Type: Primary
Antigen Symbol: GP5
Antigen Name: glycoprotein V (platelet)
Antigen Synonyms: CD42d, GPV
Clonality: Monoclonal
Clone: 508209
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR