Anti-GFP uv Mouse Monoclonal Antibody [clone: 454518]

Supplier: R & D Systems
MAB4240
220020-814EA 577.02 USD
220020-814
Anti-GFP uv Mouse Monoclonal Antibody [clone: 454518]
Antibodies
Type: Primary
Antigen Symbol: GFP uv
Antigen Name:
Antigen Synonyms: eGFP, GFPuv
Clonality: Monoclonal
Clone: 454518
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR